Please login to your online account to display your discounted pricing

AH 11110 hydrochloride, Tocris

Subtype-selective α1B ligand

$201.36 - $844.73

Chemical Identifiers

Chemical Name or Material AH 11110 hydrochloride
Assay Percent Range >98%
Molecular Formula C21H26N2O2·HCl
Formula Weight 374.91
Synonym 4-Imino-1-(2-phenylphenoxy)-4-piperidinebutan-2-ol hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

099850

 
r & d systems
RDS099850
50mg Each for $844.73

099810

 
r & d systems
RDS099810
10mg Each for $201.36
Description & Specifications

Specifications

Quantity 50mg
Formula Weight 374.91
Assay Percent Range >98%
Molecular Formula C21H26N2O2·HCl
Recommended Storage Store at Room Temperature
CAS 179388-65-9
Synonym 4-Imino-1-(2-phenylphenoxy)-4-piperidinebutan-2-ol hydrochloride
Chemical Name or Material AH 11110 hydrochloride
Safety and Handling

Store at Room Temperature