Please login to your online account to display your discounted pricing

AGN 192403 hydrochloride, Tocris


I1 selective ligand

Quantity 50mg

Manufacturer: r & d systems  RDS107250

 View more versions of this product

Catalog No. 107250

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 189.73
Quantity 50mg
Chemical Name or Material AGN 192403 hydrochloride
Assay Percent Range >98%
Molecular Formula C10H19N·HCl
CAS 1021868-90-5
Synonym (±)-2-endo-Amino-3-exo-isopropylbicyclo[2.2.1]heptane hydrochloride
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material AGN 192403 hydrochloride
Assay Percent Range >98%
Molecular Formula C10H19N·HCl
Formula Weight 189.73
Synonym (±)-2-endo-Amino-3-exo-isopropylbicyclo[2.2.1]heptane hydrochloride
Accessories

Store at Room Temperature