Please login to your online account to display your discounted pricing

AGN 192403 hydrochloride, Tocris

I1 selective ligand

$205.70 - $862.95

Chemical Identifiers

Chemical Name or Material AGN 192403 hydrochloride
Assay Percent Range >98%
Molecular Formula C10H19N·HCl
Formula Weight 189.73
Synonym (±)-2-endo-Amino-3-exo-isopropylbicyclo[2.2.1]heptane hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

107250

 
r & d systems
RDS107250
50mg Each for $862.95

107210

 
r & d systems
RDS107210
10mg Each for $205.70
Description & Specifications

Specifications

Formula Weight 189.73
Quantity 50mg
Chemical Name or Material AGN 192403 hydrochloride
Assay Percent Range >98%
Molecular Formula C10H19N·HCl
CAS 1021868-90-5
Synonym (±)-2-endo-Amino-3-exo-isopropylbicyclo[2.2.1]heptane hydrochloride
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature