missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Agmatine sulfate, 97%

Manufacturer:  Alfa Aesar™ H55363MD

 View more versions of this product

Catalog No. AAH55363MD



Description

Description

Applications
Agmatine is an intermediate in the biosynthesis of the polyamines putrescine and spermine. Agmatine binds with high affinity to α2-adrenergic receptors and imidazoline binding sites and produces blockade of the NMDA-receptor associated cation channels. Agmatine is a competitive inhibitor of nitric oxide synthase activity owing to mimicry of the natural NOS substrate arginine.

Solubility
Soluble in water, nearly insoluble in alcohol

Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifications

Specifications

Agmatine sulfate
250mg
C5H16N4O4S
MFCD00013109
14,188
Soluble in water, nearly insoluble in alcohol
C(CCN=C(N)N)CN.OS(=O)(=O)O
228.267
228.27
231°C to 238°C
2482-00-0
H2N(CH2)4NHC(=NH)NH2·H2SO4
3918807
1-4-aminobutyl guanidine sulfate
PTAYFGHRDOMJGC-UHFFFAOYSA-N
2-(4-aminobutyl)guanidine;sulfuric acid
2794990
97%

Chemical Identifiers

2482-00-0
228.267
PTAYFGHRDOMJGC-UHFFFAOYSA-N
2794990
C(CCN=C(N)N)CN.OS(=O)(=O)O
C5H16N4O4S
MFCD00013109
1-4-aminobutyl guanidine sulfate
2-(4-aminobutyl)guanidine;sulfuric acid
Safety and Handling

Safety and Handling

RTECSNumber : ME8413000

Documents