Please login to your online account to display your discounted pricing

Agarose, Standard, Low Electroendosmosis (EEO), Ultrapure Bioreagent, J.T.Baker™

High purity reagents tested for use in biotechnology applications, such as electrophoresis, and liquid chromatography

$1,610.29 - $1,610.29

Chemical Identifiers

Chemical Name or Material Agarose
CAS 9012-36-6
Name Note Standard, Low Electroendosmosis (EEO)
Purity Grade Ultrapure Bioreagent
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02002209

 
avantor performance
A42607
Poly Bottle 500g Each for $1,610.29
Description & Specifications

Specifications

Chemical Name or Material Agarose
CAS 9012-36-6
Name Note Standard, Low Electroendosmosis (EEO)
Specific Gravity 1L = 0.90kg
Packaging Poly Bottle
Purity Grade Ultrapure Bioreagent
Quantity 500g

    For Electrophoresis