missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Agarose, Low Melting Point, Analytical Grade, Promega™ DFS Item

$179.75 - $179.75

Chemical Identifiers

Physical Form Powder
Chemical Name or Material Agarose Low Melting Point
Grade Analytical Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PR-V2111 Promega Corportaion
V2111
25g Each for $179.75
Specifications Description & Specifications

Chemical Identifiers

Powder
Agarose Low Melting Point
Analytical Grade

Specifications

Powder
25g
Analytical Grade
White
Agarose Low Melting Point

Safety and Handling

Recommended Storage : Store at 22° to 25°C.