missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Agarose (Low-EEO/Multi-Purpose/Molecular Biology Grade), Fisher BioReagents

Manufacturer:  Fisher Scientific Company BP160500

 View more versions of this product

Catalog No. BP160-500



 

Specifications Description & Specifications

Specifications

Molecular Biology Grade
500g
C12H18O9
Light Cream
DNase- and RNase-Free
9012-36-6
0.09 to 0.13
0.15% max.
Agarose
Powder/Solid
1200g/cm2min.
DNase free
Pass Test
Poly Bottle
Low-EEO/Multi-Purpose
34.5° to 37.5°C
Pass Test

Chemical Identifiers

9012-36-6
C12H18O9
Molecular Biology Grade, DNase- and RNase-Free
Powder/Solid
Agarose, Low-EEO/Multi-Purpose

Safety and Handling

SDS & Safety Documents

Recommended Storage : Room temperature

Promotions

Promotions

  • When you buy two bottles of Fisher BioReagents chemicals, you’ll get a third bottle at no additional cost. Or, buy four bottles of chemicals and get two bottles at no additional cost.

    09/30/17