Please login to your online account to display your discounted pricing

Agarose (Isoelectric Focusing of Proteins/IEF Grade), Fisher BioReagents


Quantity 10g
Packaging Poly Bottle

Manufacturer: fisher scientific company  FLBP16310

 View more versions of this product

Catalog No. BP163-10

  • / Each
  • / Case of 6 Each
 

Description & Specifications

Chemical Identifiers

Physical Form Powder/Solid
Formula Weight 306.12
Quantity 10g
Gel Strength 500g/cm2min.
DNase DNase free
Purity Grade IEF Grade
Molecular Formula C12H18O9
Color Light Cream
Chemical Name or Material Agarose
Electrophoresis Pass Test
Electroendosmosis (EEO) Not Detected
Gelation Temperature 34° to 45°C
Identification Pass Test
Name Note Isoelectric Focusing of Proteins
Packaging Poly Bottle
CAS 9012-36-6
ChemAlert Storage Symbol Gray
Sulfate 0.20% max.
Recommended Storage RT

Specifications

Chemical Name or Material Agarose
Name Note Isoelectric Focusing of Proteins
CAS 9012-36-6
Purity Grade IEF Grade
Molecular Formula C12H18O9
Formula Weight 306.12
Accessories
 

RT

SDS & Safety Documents