missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Agarose (Isoelectric Focusing of Proteins/IEF Grade), Fisher BioReagents

$232.76 - $232.76

Chemical Identifiers

CAS 9012-36-6
Molecular Formula C12H18O9
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
BP163-10 SDS View Documents Fisher BioReagents
FLBP16310
10g Poly Bottle Each for $232.76
Case of 6 Each for $1,396.56
 
Description

Description

Highly purified agarose for separating proteins by isoelectric focusing.
Specifications

Chemical Identifiers

9012-36-6
C12H18O9

Specifications

9012-36-6
Pass Test
34° to 45°C
Pass Test
Poly Bottle
306.12
EIF
10g
DNase free
Not Detected
500g/cm2min.
C12H18O9
0.20% max.
Powder/Solid
Cream
Agarose, Isoelectric Focusing of Proteins
Safety and Handling

Safety and Handling

Recommended Storage : RT