Please login to your online account to display your discounted pricing

Agarose, High Resolution, Molecular Biology Reagent, MP Biomedicals


Quantity 100g

Manufacturer: mp biomedicals  19398480

 View more versions of this product

Catalog No. ICN19398480

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 100g
CAS 9012-36-6
Name Note High Resolution, Molecular Biology Reagent
Chemical Name or Material Agarose
MDL Number MFCD00081294
Synonym 3, 6-Anhydro-α-L-galacto-β-D-galactan
Recommended Storage Room Temperature

Specifications

Chemical Name or Material Agarose
Name Note High Resolution, Molecular Biology Reagent
CAS 9012-36-6
MDL Number MFCD00081294
Synonym 3, 6-Anhydro-α-L-galacto-β-D-galactan
Accessories

Room Temperature