Please login to your online account to display your discounted pricing

Agarose, High Resolution, Molecular Biology Reagent, MP Biomedicals

$517.75 - $517.75

Chemical Identifiers

Chemical Name or Material Agarose
Name Note High Resolution, Molecular Biology Reagent
CAS 9012-36-6
MDL Number MFCD00081294
Synonym 3, 6-Anhydro-α-L-galacto-β-D-galactan
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

ICN19398480

 
mp biomedicals
19398480
100g Each for $517.75
Description & Specifications

Specifications

Quantity 100g
Chemical Name or Material Agarose
Name Note High Resolution, Molecular Biology Reagent
MDL Number MFCD00081294
CAS 9012-36-6
Synonym 3, 6-Anhydro-α-L-galacto-β-D-galactan
Recommended Storage Room Temperature