Please login to your online account to display your discounted pricing

Agarose, Genetic Technology Grade, MP Biomedicals™


Quantity 125g

Manufacturer: mp biomedicals  04800568

 View more versions of this product

Catalog No. ICN800568

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 125g
Chemical Name or Material Agarose
CAS 9012-36-6
Purity Grade Genetic Technology Grade

Specifications

Chemical Name or Material Agarose
CAS 9012-36-6
Purity Grade Genetic Technology Grade