missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Agarose gel loading dye (6X), alkaline

$89.45 - $89.45

Specifications

Chemical Name or Material Agarose gel loading dye
Name Note alkaline
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ62157AB View Documents Alfa Aesar™
J62157AB
10mL Each for $89.45
 
AAJ62157AC View Documents Alfa Aesar™
J62157AC
25mL N/A N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Applications
Agarose gel loading dye (6X), alkaline is used in DNA gel analysis in alkaline condition. It is used in agarose gel electrophoresis.

Solubility
Fully miscible in water.

Notes
300mM sodium hydroxide, 6mM EDTA, 0.25% bromocresol green, 0.25% xylene cyanol FF and 18% Ficoll
Specifications

Specifications

Agarose gel loading dye
alkaline