missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Agarose Gel Loading Dye (6X, Alkaline)

Manufacturer:  Thermo Scientific™ J62157AC

Catalog No. AAJ62157AC

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Agarose gel loading dye (6X), alkaline is used in DNA gel analysis in alkaline condition. It is used in agarose gel electrophoresis.

Solubility
Fully miscible in water.

Notes
300mM sodium hydroxide, 6mM EDTA, 0.25% bromocresol green, 0.25% xylene cyanol FF and 18% Ficoll
Specifications

Specifications

Agarose gel loading dye
6X
Fully miscible in water.
alkaline
25mL
Liquid
SDS
Documents
Product Certifications
Promotions

Promotions

RUO – Research Use Only