missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Agar Agar, BAKER™, J.T.Baker™

A grade of chemicals for laboratory use

Manufacturer:  J.T.Baker A43405

Catalog No. 2002204



Specifications

Specifications

Agar Agar
100g
C14H24O9
GYYDPBCUIJTIBM-DYOGSRDZSA-N
(2R,3S,4S,5R)-2-(hydroxymethyl)-6-[[(4R,5S)-4-hydroxy-3-methyl-2,6-dioxabicyclo[3.2.1]octan-8-yl]oxy]-4-methoxyoxane-3,5-diol
71571511
Poly Bottle
9002-18-0
3r,4s,5s,6r-2-4r,5s-4-hydroxy-3-methyl-2,6-dioxabicyclo 3.2.1 octan-8-yl oxy-6-hydroxymethyl-4-methoxyoxane-3,5-diol
CC1C(C2C(C(O1)CO2)OC3C(C(C(C(O3)CO)O)OC)O)O
336.337

Chemical Identifiers

9002-18-0
336.337
3r,4s,5s,6r-2-4r,5s-4-hydroxy-3-methyl-2,6-dioxabicyclo 3.2.1 octan-8-yl oxy-6-hydroxymethyl-4-methoxyoxane-3,5-diol
(2R,3S,4S,5R)-2-(hydroxymethyl)-6-[[(4R,5S)-4-hydroxy-3-methyl-2,6-dioxabicyclo[3.2.1]octan-8-yl]oxy]-4-methoxyoxane-3,5-diol
C14H24O9
GYYDPBCUIJTIBM-DYOGSRDZSA-N
71571511
CC1C(C2C(C(O1)CO2)OC3C(C(C(C(O3)CO)O)OC)O)O
Documents