Please login to your online account to display your discounted pricing

AG 99, Tocris

EGFR-kinase inhibitor

$159.50 - $669.90

Chemical Identifiers

Chemical Name or Material AG 99
Assay Percent Range >99%
Molecular Formula C10H8N2O3
Formula Weight 204.18
Synonym (E)-2-Cyano-3-(3, 4-dihydroxyphenyl)-2-propenamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

049750

 
r & d systems
RDS049750
50mg Each for $669.90

049710

 
r & d systems
RDS049710
10mg Each for $159.50
Description & Specifications

Specifications

Quantity 50mg
Formula Weight 204.18
Chemical Name or Material AG 99
Assay Percent Range >99%
Molecular Formula C10H8N2O3
CAS 122520-85-8
Synonym (E)-2-Cyano-3-(3, 4-dihydroxyphenyl)-2-propenamide
Recommended Storage Desiccate at +4°C
Safety and Handling

Desiccate at +4°C