missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

AG 825, Tocris

Selective ErbB2 inhibitor

$166.60 - $693.49

Chemical Identifiers

Assay Percent Range >98%
Molecular Formula C19H15N3O3S2
Formula Weight 397.47
Synonym (E)-3-[3-[2-Benzothiazolythio)methyl]-4-hydroxy-5-methoxyphenyl]-2-cyano-2-propenamide
Chemical Name or Material AG 825
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
155510 R & D Systems
RDS155510
10mg Each for $166.60
155550 R & D Systems
RDS155550
50mg Each for $693.49
Specifications Description & Specifications

Chemical Identifiers

>98%
397.47
AG 825
C19H15N3O3S2
(E)-3-[3-[2-Benzothiazolythio)methyl]-4-hydroxy-5-methoxyphenyl]-2-cyano-2-propenamide

Specifications

149092-50-2
C19H15N3O3S2
(E)-3-[3-[2-Benzothiazolythio)methyl]-4-hydroxy-5-methoxyphenyl]-2-cyano-2-propenamide
AG 825
>98%
397.47
10mg

Safety and Handling

Recommended Storage : Store at +4°C