Please login to your online account to display your discounted pricing

AG 556, Tocris


EGFR-kinase inhibitor

Quantity 10mg

Manufacturer: r & d systems  RDS061610

 View more versions of this product

Catalog No. 061610

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 336.39
CAS 133550-41-1
Chemical Name or Material AG 556
Assay Percent Range >99%
Molecular Formula C20H20N2O3
Synonym (E)-2-Cyano-3-(3, 4-dihydroxyphenyl)-N-(4-phenylbutyl)-2-propenamide
Recommended Storage Desiccate at +4°C

Specifications

Chemical Name or Material AG 556
Assay Percent Range >99%
Molecular Formula C20H20N2O3
Formula Weight 336.39
Synonym (E)-2-Cyano-3-(3, 4-dihydroxyphenyl)-N-(4-phenylbutyl)-2-propenamide
Accessories

Desiccate at +4°C