Please login to your online account to display your discounted pricing

AG 556, Tocris

EGFR-kinase inhibitor

$168.15 - $697.40

Chemical Identifiers

Chemical Name or Material AG 556
Assay Percent Range >99%
Molecular Formula C20H20N2O3
Formula Weight 336.39
Synonym (E)-2-Cyano-3-(3, 4-dihydroxyphenyl)-N-(4-phenylbutyl)-2-propenamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

061650

 
r & d systems
RDS061650
50mg Each for $697.40

061610

 
r & d systems
RDS061610
10mg Each for $168.15
Description & Specifications

Specifications

Quantity 50mg
Formula Weight 336.39
Chemical Name or Material AG 556
Assay Percent Range >99%
Molecular Formula C20H20N2O3
CAS 133550-41-1
Synonym (E)-2-Cyano-3-(3, 4-dihydroxyphenyl)-N-(4-phenylbutyl)-2-propenamide
Recommended Storage Desiccate at +4°C
Safety and Handling

Desiccate at +4°C