Please login to your online account to display your discounted pricing

AG 555, Tocris

Potent EGFR-kinase inhibitor

$164.61 - $682.67

Chemical Identifiers

Chemical Name or Material AG 555
Assay Percent Range >99%
Molecular Formula C19H18N2O3
Formula Weight 322.36
Synonym (E)-2-Cyano-3-(3, 4-dihydroxyphenyl)-N-(3-phenylpropyl)-2-propenamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

061810

 
r & d systems
RDS061810
10mg Each for $164.61

061850

 
r & d systems
RDS061850
50mg Each for $682.67
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 322.36
Chemical Name or Material AG 555
CAS 133550-34-2
Molecular Formula C19H18N2O3
Synonym (E)-2-Cyano-3-(3, 4-dihydroxyphenyl)-N-(3-phenylpropyl)-2-propenamide
Assay Percent Range >99%
Recommended Storage Desiccate at +4°C
Safety and Handling

Desiccate at +4°C