Please login to your online account to display your discounted pricing

AG 494, Tocris

Potent EGFR-kinase inhibitor

$159.50 - $678.85

Chemical Identifiers

Chemical Name or Material AG 494
Assay Percent Range >98%
Molecular Formula C16H12N2O3
Formula Weight 280.28
Synonym (E)-2-Cyano-3-(3, 4-dihydroxyphenyl)-N-phenyl-2-propenamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

061910

 
r & d systems
RDS061910
10mg Each for $159.50

061950

 
r & d systems
RDS061950
50mg Each for $678.85
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 280.28
Assay Percent Range >98%
Recommended Storage Desiccate at +4°C
Molecular Formula C16H12N2O3
Chemical Name or Material AG 494
Synonym (E)-2-Cyano-3-(3, 4-dihydroxyphenyl)-N-phenyl-2-propenamide
CAS 133550-35-3
Safety and Handling

Desiccate at +4°C