Please login to your online account to display your discounted pricing

AG 494, Tocris


Potent EGFR-kinase inhibitor

Quantity 50mg

Manufacturer: r & d systems  RDS061950

 View more versions of this product

Catalog No. 061950

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 50mg
Formula Weight 280.28
Chemical Name or Material AG 494
Assay Percent Range >98%
Molecular Formula C16H12N2O3
CAS 133550-35-3
Synonym (E)-2-Cyano-3-(3, 4-dihydroxyphenyl)-N-phenyl-2-propenamide
Recommended Storage Desiccate at +4°C

Specifications

Chemical Name or Material AG 494
Assay Percent Range >98%
Molecular Formula C16H12N2O3
Formula Weight 280.28
Synonym (E)-2-Cyano-3-(3, 4-dihydroxyphenyl)-N-phenyl-2-propenamide
Accessories

Desiccate at +4°C