Please login to your online account to display your discounted pricing

AG 18, Tocris


EGFR/PDGFR-kinase inhibitor

Quantity 10mg

Manufacturer: r & d systems  RDS049310

 View more versions of this product

Catalog No. 049310

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 10mg
Formula Weight 186.17
Chemical Name or Material AG 18
Assay Percent Range >98%
Molecular Formula C10H6N2O2
CAS 118409-57-7
Synonym [(3, 4-Dihydroxyphenyl)methylene]-propenedinitrile
Recommended Storage Desiccate at +4°C

Specifications

Chemical Name or Material AG 18
Assay Percent Range >98%
Molecular Formula C10H6N2O2
Formula Weight 186.17
Synonym [(3, 4-Dihydroxyphenyl)methylene]-propenedinitrile
Accessories

Desiccate at +4°C