Please login to your online account to display your discounted pricing

AG 18, Tocris

EGFR/PDGFR-kinase inhibitor

$162.94 - $684.37

Chemical Identifiers

Chemical Name or Material AG 18
Assay Percent Range >98%
Molecular Formula C10H6N2O2
Formula Weight 186.17
Synonym [(3, 4-Dihydroxyphenyl)methylene]-propenedinitrile
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

049350

 
r & d systems
RDS049350
50mg Each for $684.37

049310

 
r & d systems
RDS049310
10mg Each for $162.94
Description & Specifications

Specifications

Quantity 50mg
Formula Weight 186.17
Chemical Name or Material AG 18
Assay Percent Range >98%
Molecular Formula C10H6N2O2
CAS 118409-57-7
Synonym [(3, 4-Dihydroxyphenyl)methylene]-propenedinitrile
Recommended Storage Desiccate at +4°C
Safety and Handling

Desiccate at +4°C