Please login to your online account to display your discounted pricing

AFP Mouse anti-Human, Clone: 1E8, eBioscience™

Mouse Monoclonal Antibody

$205.90 - $433.55

Specifications

Antigen AFP
Clone 1.00E+08
Host Species Murine
Gene Alias AFP
Species Reactivity Human
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501122787

 
life technologies
LS14949980
25μg Each for $205.90

501122788

 
life technologies
LS14949982
100μg Each for $433.55
Description & Specifications

Specifications

Antigen AFP
Clone 1.00E+08
Host Species Murine
Gene Alias AFP
Species Reactivity Human
Applications Immunocytochemistry
Applications Immunohistochemistry (Formalin/Paraffin)
Applications Western Blot
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.5mg/mL

This 1E8 monoclonal antibody reacts with human alpha-fetoprotein (AFP). This 70kDa secretory protein is a member of the albumin gene family. Synthesized by the yolk sac and fetal liver during embryogenesis, AFP protein levels are highest in fetal serum. After birth, serum AFP levels decrease dramatically. In fact, AFP is nearly undetectable in normal adult serum. However, hepatocellular carcinoma and germ cell teratoblastoma, as well as liver regeneration, viral hepatitis, and cirrhosis, leads to elevated AFP serum levels in adults. As such, detection of this protein is frequently used as a diagnostic marker for these conditions.