missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Applied Biosystems™ ExoSAP-IT™ For PCR Product Clean-Up
Designed for simple, quick PCR cleanup for downstream applications, such as DNA sequencing or Single Nucleotide Polymorphism (SNP) analysis. Single-step enzymatic cleanup of PCR products that eliminates unincorporated primers and dNTPs.
$171.00 - $5,380.00
Specifications
For Use With (Application) | Quick PCR clean-up for downstream applications; Eliminates unincorporated primers and dNTPs |
---|---|
Storage Requirements | −20°C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AF782011
|
Applied Biosystems™
LS782011ML |
1mL |
Each for $660.00
|
|
AF782024X1
|
Applied Biosystems™
LS782024X1ML |
4 × 1mL |
Each for $2,375.00
|
|
AF7820510
|
Applied Biosystems™
LS7820510ML |
10mL |
Each for $5,380.00
|
|
AF78200200
|
Applied Biosystems™
LS78200200UL |
200μL |
Each for $171.00
|
|
Description
- Two hydrolytic enzymes, Exonuclease I and Shrimp Alkaline Phosphatase (SAP), together in a specially formulated buffer
- 30-minute protocol
Specifications
Quick PCR clean-up for downstream applications; Eliminates unincorporated primers and dNTPs | |
−20°C |