Please login to your online account to display your discounted pricing

ExoSAP-IT™ For PCR Product Clean-Up


Designed for simple, quick PCR cleanup for downstream applications, such as DNA sequencing or Single Nucleotide Polymorphism (SNP) analysis. Single-step enzymatic cleanup of PCR products that eliminates unincorporated primers and dNTPs.

Manufacturer: life technologies  LS782024X1ML

 View more versions of this product

Catalog No. AF782024X1

  • / Each
 

Description & Specifications

Specifications

Quantity 4 × 1mL
No. of Reactions 2,000 reactions
Storage Requirements −20°C
For Use With (Application) Quick PCR clean-up for downstream applications; Eliminates unincorporated primers and dNTPs
Product Type Single-step enzymatic PCR Cleanup Kit
Product Name Single-step enzymatic PCR Cleanup Kit
Quantity 2000 reactions

  • Two hydrolytic enzymes, Exonuclease I and Shrimp Alkaline Phosphatase (SAP), together in a specially formulated buffer
  • 30-minute protocol