Please login to your online account to display your discounted pricing
Affymetrix™ Exonuclease I
Useful in preparing PCR products for applications involving sequencing or labeling methods
Manufacturer: life technologies  LS70073X5000UN
 View more versions of this product
 Disclaimers
The purchase of ExoSAP-IT provides a license to the methods of PCR clean up using Exonuclease I and SAP.
Catalog No. AF70073X500
  • / Each
Description & Specifications
Specifications
Product Name Exonuclease I
Quantity 5,000U
Concentration 10U/μL
Quantity 5,000U
Concentration 10U/μL