Please login to your online account to display your discounted pricing

AF-DX 116, Tocris

Selective M2 antagonist

$199.70 - $834.54

Chemical Identifiers

Chemical Name or Material AF-DX 116
Assay Percent Range >98%
Molecular Formula C24H31N5O2
Formula Weight 421.54
Synonym 11-[[2-[(Diethylamino)methyl]-1-piperidinyl]acetyl]-5,11-dihydro-6H-pyrido[2,3-b][1,4]benzodiazepin-6-one
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

110510

 
r & d systems
110510
10mg Each for $199.70

110550

 
r & d systems
110550
50mg Each for $834.54
Description & Specifications

Specifications

Formula Weight 421.54
Quantity 10mg
Chemical Name or Material AF-DX 116
Molecular Formula C24H31N5O2
Assay Percent Range >98%
CAS 102394-31-0
Synonym 11-[[2-[(Diethylamino)methyl]-1-piperidinyl]acetyl]-5,11-dihydro-6H-pyrido[2,3-b][1,4]benzodiazepin-6-one
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature