AF-DX 116, Tocris
Selective M2 antagonist
Manufacturer:  R & D Systems 110510
 View more versions of this product
Catalog No. 110510
Specifications Description & Specifications
Specifications
AF-DX 116
>98%
C24H31N5O2
11-[[2-[(Diethylamino)methyl]-1-piperidinyl]acetyl]-5,11-dihydro-6H-pyrido[2,3-b][1,4]benzodiazepin-6-one
102394-31-0
10mg
421.54
Chemical Identifiers
>98%
421.54
AF-DX 116
C24H31N5O2
11-[[2-[(Diethylamino)methyl]-1-piperidinyl]acetyl]-5,11-dihydro-6H-pyrido[2,3-b][1,4]benzodiazepin-6-one
Safety and Handling
Recommended Storage : Store at Room Temperature