Aerosol™ OT Solution, 10% (w/w), Fisher Chemical
Aerosol™ OT 10% is a remarkable chemical solution that “makes water wetter” because it reduces surface tension and interfacial tension to a considerable degree even at low concentrations
Manufacturer:  Fisher Chemical SA2924
Catalog No. SA292-4
Description
Description
Specifications
Specifications
Aerosol™ OT Solution, 10% (w/w)
10% (w/w)
9 to 11%
Pass Test
Glass Bottle
Contains: Sodium Dioctyl Sulfosuccinate,Butyl Cellosolve™,Water
4L
10% w/w
577-11-7,111-76-2,7732-18-5,64-17-5,104-76-7
10% w/w
MFCD00012455
Liquid
Undesignated
SDS
Documents
Accessories