Axygen™ Aerosol Filter Tips, Non-sterile
Prevent cross-contamination of samples. Axygen™ Aerosol Filter Tips, Non-sterile feature a universal fit. All tips comply with USP 3127 (Heavy Metal Contamination) and are free of leachable additives. Compatible with a variety of pipettes and available in a range of filter sizes.
$139.32 - $176.99
Specifications
Certifications/Compliance DNase, RNase, pyrogen free
USP 3127 (Heavy Metal Contamination).
Material Polypropylene
Product Type Aerosol Barrier Tip
Color Clear
Sterility Nonsterile
View More Specs
Products ${productFamilyLength}
Catalog Number Mfr. No. Tip Style Packaging Volume (Metric) Price Quantity  
Catalog Number Mfr. No. Tip Style Packaging Volume (Metric) Price Quantity  
14-222-764 Axygen™
TF1000
Barrier, Universal Fit Bulk: 1000 Tips/Pk., 5 Packs/Cs. 100 to 1000μL Pack of 1000 Each for $176.99
Case of 5 Pack for $769.50
14-222-775 Axygen™
TF20
Barrier, Universal Fit Bulk 0.5 to 20μL Pack of 1000 Each for $160.56
Case of 10 Pack for $1,396.20
14-222-776 Axygen™
TF200
Barrier, Universal Fit Bulk 1 to 200μL Pack of 1000 Each for $154.87
Case of 10 Pack for $1,346.70
14-222-786 Axygen™
TF350
Barrier, Universal Fit Bulk: 1000 Tips/Pk/, 10 Packs/Cs. 1 to 300μL Pack of 1000 Each for $139.32
Case of 10 Pack for $1,211.50
Specifications
Description
Technologically advanced filter barrier pipette tips
  • Ideal for DNA amplification and microbiological applications where trace contamination can invalidate results
  • Filter allows free passage of air while preventing cross-contamination by blocking the passage of aerosols or liquids
  • Nonpyrogenic, clear polyethylene filters are free from DNA inhibitors and leachable additives
  • Every filter is inserted into the tip robotically to ensure freedom from nuclease contamination
  • Packaged in sealed-bottom racks with lids to prevent contamination
Order Info
Available in bulk or racked formats.
Certifications
Certified RNase-, DNase- and Pyrogen free
Description & Specifications
Specifications
DNase, RNase, pyrogen free
USP 3127 (Heavy Metal Contamination).
Aerosol Barrier Tip
Nonsterile
Polypropylene
Clear