Please login to your online account to display your discounted pricing

AEBSF Hydrochloride (White to Off-white Crystals), Fisher BioReagents


Quantity 100mg
Packaging Poly Micro Tube

Manufacturer: fisher scientific company  FLBP635100

 View more versions of this product

Catalog No. BP635-100

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 239.7
Physical Form Powder/Solid
Melting Point 175°C
Quantity 100mg
TSCA Not on TSCA inventory: for R and D use only; not for manufacturing or commercial purposes.
Molecular Formula C8H10FNO2S · HCl
Color White
Packaging Poly Micro Tube
Assay Percent Range ≥98.0 %
Chemical Name or Material AEBSF Hydrochloride
Identification Pass Test
Hydrogen (H) 4.58 to 4.66%
Nitrogen (N) 5.64 to 6.04%
Purity ≥98 (HPLC)
CAS 30827-99-7
ChemAlert Storage Symbol Gray
Solubility in Alcohol Pass Test
Synonym 4-(2-Aminoethyl)-Benzenesulfonyl Fluoride HCl
Sulfur (S) 12.98 to 13.78%
Health Hazard 1 Signal Word:DANGER!
Health Hazard 3 Emergency Overview
Corrosive. Irritating to eyes and skin. May cause burns.
NFPA
Health: 3
Flammability:
Carbon (C) 39.59 to 40.59%
Recommended Storage RT

Specifications

Chemical Name or Material AEBSF Hydrochloride
CAS 30827-99-7
Physical Form Powder/Solid
Assay Percent Range ≥98.0 %
Molecular Formula C8H10FNO2S · HCl
Formula Weight 239.7
Synonym 4-(2-Aminoethyl)-Benzenesulfonyl Fluoride HCl
Accessories
 

Signal Word:DANGER!

Emergency Overview
Corrosive. Irritating to eyes and skin. May cause burns.
NFPA
Health: 3
Flammability:

RT

SDS & Safety Documents