Please login to your online account to display your discounted pricing

AEBSF Hydrochloride (White to Off-white Crystals), Fisher BioReagents

$205.66 - $676.17

Chemical Identifiers

Chemical Name or Material AEBSF Hydrochloride
CAS 30827-99-7
Physical Form Powder/Solid
Assay Percent Range ≥98.0 %
Molecular Formula C8H10FNO2S · HCl
Formula Weight 239.7
Synonym 4-(2-Aminoethyl)-Benzenesulfonyl Fluoride HCl
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

BP635-100

  SDS
fisher scientific company
FLBP635100
Poly Micro Tube 100mg Each for $229.44

BP635-50

  SDS
fisher scientific company
FLBP63550
Poly Micro Tube 50mg Each for $205.66

BP635-500

  SDS
fisher scientific company
FLBP635500
Poly Micro Tube 500mg Each for $676.17
Description & Specifications

Specifications

Formula Weight 239.7
Physical Form Powder/Solid
Melting Point 175°C
Quantity 100mg
TSCA Not on TSCA inventory: for R and D use only; not for manufacturing or commercial purposes.
Molecular Formula C8H10FNO2S · HCl
Color White
Packaging Poly Micro Tube
Assay Percent Range ≥98.0 %
Chemical Name or Material AEBSF Hydrochloride
Identification Pass Test
Hydrogen (H) 4.58 to 4.66%
Nitrogen (N) 5.64 to 6.04%
Purity ≥98 (HPLC)
CAS 30827-99-7
ChemAlert Storage Symbol Gray
Solubility in Alcohol Pass Test
Synonym 4-(2-Aminoethyl)-Benzenesulfonyl Fluoride HCl
Sulfur (S) 12.98 to 13.78%
Health Hazard 1 Signal Word:DANGER!
Health Hazard 3 Emergency Overview
Corrosive. Irritating to eyes and skin. May cause burns.
NFPA
Health: 3
Flammability:
Carbon (C) 39.59 to 40.59%
Recommended Storage RT

This specific, irreversible inhibitor of serine proteases is a stable, nontoxic alternative to PMSF.

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling
 

Signal Word:DANGER!

Emergency Overview
Corrosive. Irritating to eyes and skin. May cause burns.
NFPA
Health: 3
Flammability:

RT