missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

AEBSF Hydrochloride (White to Off-white Crystals), Fisher BioReagents

Manufacturer:  Fisher Scientific Company FLBP635500

 View more versions of this product

Catalog No. BP635-500


 

Specifications Description & Specifications

Specifications

AEBSF Hydrochloride
39.59 to 40.59%
4.58 to 4.66%
Poly Micro Tube
≥98.0 %
C8H10FNO2S · HCl
Pass Test
12.98 to 13.78%
175°C
30827-99-7
Pass Test
5.64 to 6.04%
Powder/Solid
≥98 (HPLC)
239.7
4-(2-Aminoethyl)-Benzenesulfonyl Fluoride HCl
White
500mg

Chemical Identifiers

30827-99-7
≥98.0 %
239.7
AEBSF Hydrochloride
Powder/Solid
C8H10FNO2S · HCl
4-(2-Aminoethyl)-Benzenesulfonyl Fluoride HCl

Safety and Handling

SDS & Safety Documents

Emergency Overview
Corrosive. Irritating to eyes and skin. May cause burns.
NFPA
Health: 3
Flammability:

Signal Word:DANGER!

TSCA : Not on TSCA inventory: for R and D use only; not for manufacturing or commercial purposes.

Recommended Storage : RT