Please login to your online account to display your discounted pricing

ADX 10059 hydrochloride, Tocris™

Negative allosteric modulator at mGlu5

$239.63 - $976.15

Chemical Identifiers

Chemical Name or Material ADX 10059 hydrochloride
Assay Percent Range >99%
Molecular Formula C15H13FN2·HCl
Formula Weight 276.74
Synonym 2-[(3-Fluorophenyl)ethynyl]-4, 6-dimethyl-3-pyridinamine hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

441610

 
r & d systems
RDS441610
10mg Each for $239.63

441650

 
r & d systems
RDS441650
50mg Each for $976.15
Description & Specifications

Specifications

Formula Weight 276.74
Quantity 10mg
Chemical Name or Material ADX 10059 hydrochloride
Assay Percent Range >99%
Molecular Formula C15H13FN2·HCl
CAS 757949-98-7
Synonym 2-[(3-Fluorophenyl)ethynyl]-4, 6-dimethyl-3-pyridinamine hydrochloride
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature