missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Ohaus™ ADVENTURER™ PRECISION

Ready For Your Lab, Wherever That May Be.

Manufacturer:  Ohaus™ OHS30100632RM

 View more versions of this product

Catalog No. 01-919-365



Description

Description

Striking the ideal balance between inventive features and functional, uncomplicated weighing capabilities, the OHAUS Adventurer incorporates all of the applications necessary for routine weighing and measurement activities. -With a color touchscreen, GLP/GMP compliance capabilities, two USB ports, and much more, Adventurer is the most complete balance in its class.

Warranty

2 Year

Specifications

Specifications

AC Adapter (Included)
NTEP
340mm
230mm
Cast Aluminun base
5.8kg
50/60 Hz
Touchscreen LCD
AC
0.001g
Draftshield
AX423N/E
2s
354mm
420g
420g
Documents