Please login to your online account to display your discounted pricing
Advantech Replacement Parts for Model L3P Sonic Sifter Separators
$46.60 - $1,520.58
Specifications
For Use With (Equipment) L3P sonic sifter separators
View More Specs
Products
Catalog Number Mfr. No. Product Type Price Quantity    
14-294-18
advantech manufacturing
ATML3N3
Top Cone Each for $46.60
14-294-16
advantech manufacturing
ATML3N1
Column Lock Each for $673.25
14-294-17
advantech manufacturing
L3-N2
Diaphragm Each for $78.04
14-294-20
advantech manufacturing
L3-N5
Fines Collector Each for $175.79
Pack of 10 Each for $1,757.90
14-294-22
advantech manufacturing
ATML3N7
Complete stack assembly Each for $1,520.58
14-294-21
advantech manufacturing
L3-N6
Fines collector holder Each for $834.47
Description & Specifications
Specifications
For Use With (Equipment) L3P sonic sifter separators
Five components, sold separately or as complete assembly.
  • Column Lock holds column taut
  • Dust Stop/Diaphragm transmits oscillary motion to air column and keeps particles within column
  • Top Cone sits atop spacers
  • Fines Collector stops dust at bottom of air column in sifter and transmits oscillary motion to air column
  • Fines Collector Holder supports column
  • Complete Stack Assembly includes all components described above, plus six standard spacers (14-294-19).