MSA™ Advantage™ 4000 Twin-Port Facepiece
Two bayonet cartridge adapters mount directly onto the facepiece lens for compatibility with all Advantage cartridges and filters
$476.78 - $519.37
Specifications
Product Type | Twin-Port Facepiece |
---|---|
For Use With (Equipment) | Msa™ respirator models |
Catalog Number | Mfr. No. | Material | Size | Harness Style | Price | Quantity | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Material | Size | Harness Style | Price | Quantity | |||
17-200-090
|
MSA™
10108570 |
Rubber | Small | Dupont™ Kevlar™ aramid fiber net |
Each for $519.37
|
|
17-200-088
|
MSA™
10108568 |
Rubber | Medium | Dupont™ Kevlar™ aramid fiber net |
Each for $519.37
|
|
17-200-092
|
MSA™
10108572 |
Rubber | Large | Dupont™ Kevlar™ aramid fiber net |
Each for $519.37
|
|
17-200-089
|
MSA™
10108569 |
Silicone | Small | Dupont™ Kevlar™ aramid fiber net |
Each for $519.37
|
|
17-200-087
|
MSA™
10108567 |
Silicone, Rubber | Medium | Kevlar net, four-point |
Each for $519.37
|
|
17-200-091
|
MSA™
10108571 |
Silicone | Large | Dupont™ Kevlar™ aramid fiber net |
Each for $519.37
|
|
18-139-431
|
MSA™
10083783 |
Rubber | Medium | Adjustable silicone rubber |
Each for $476.78
|
|
Description
- Approved with all MSA Advantage Cartridges
- Features two mounting locations for cartridges or filters on the right and left sides
- Standard nose cup with twin inhalation check valves
Compliance
NIOSH approved under 42 CFR 84 for use against a variety of respiratory hazards when used as designed with appropriate cartridges and filters in conjunction with OSHA fit testing requirement.
Specifications
Twin-Port Facepiece | |
Msa™ respirator models |