Advanced Snap Circuits Kit
Advanced topics challenge knowledge and creativity.
Manufacturer: Fisher Science Education SC500R
Requires : Four AA batteries (S43921)
Description
More than 75 modular parts easily and safely build over 500 experiments. Designed for high school and vocational training, this advanced electronics kit explores the technology behind AM/FM radios, digital recording, seven-segment LED displays and more.
- Included student manual relates information about topics such as resistors, capacitors, transistors, motors, various types of circuits, diodes, relays and transformers
- Parts are stored in a lightweight, durable case with custom foam inserts
- Teaching guide prepares educators for proper instruction and includes quizzes
Order Info
Student and teaching guides help integrate more advanced topics in electronics. Optional computer interface takes learning to the next level with an additional 73 experiments.
Specifications
Snap Circuits™ Educational Series | |
500 projects, student and teacher guides and custom storage case | |
8.4 lb. | |
3.81 kg |
High school and vocational training, this advanced electronics kit explores the technology behind AM/FM radios, digital recording, seven-segment LED displays and more | |
7.75 lb. | |
3.5 kg |