missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3M™ Advanced Protection Disposable Face Masks
$37.32 - $37.61
Specifications
Color | Clear |
---|---|
Product Type | Safety System |
For Use With (Application) | Provides impact protection |
Includes | Combination of ratchet headgear and WP96 Face Shield; Five position crown strap |
Catalog Number | Mfr. No. | Material | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Material | Price | Quantity | |||||
19-145-456
|
3M™
7000127244 |
Polycarbonate |
Each for $37.61
Case of 5 Each for $188.05 |
|
19-145-465
|
3M™
7000127243 |
Propionate |
Each for $37.32
Case of 5 Each for $186.60 |
|
Description
- 3M Catalog# 82782-00000 , 3M Catalog# 82783-00000
Specifications
Clear | |
Safety System | |
Provides impact protection | |
Combination of ratchet headgear and WP96 Face Shield; Five position crown strap |