3M™ Ratchet Headgear H8A with Clear WP96 Polycarbonate Faceshield
3M™ ratchet head gear with 3M™ polycarbonate face shield provides head and face protection in working environments with sparks, chips and debris. This safety system includes ratchet head gear and a 3M™ polycarbonate face shield.
Supplier:  3M™ 7000127244
Catalog No. 19-145-456
Description
Description
  • Five-position crown strap 
  • Offers face protection for many jobs and tasks where liquid and chemical splashes may occur
  • Safety system comes with headgear and faceshield together
  • 3M Catalog# 82701-00000 , 3M Catalog# 82783-00000
Specifications
Specifications
9 x 14.25 in.
Safety System
Combination of ratchet headgear and WP96 face shield; Five position crown strap
Polycarbonate faceshield
Clear
Provides impact protection
82783-00000