missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Advanced Instruments Model 3250 Single-Sample Osmometer

Provides fast, accurate test results using 200–250μL

Manufacturer:  Advanced Instruments AI3250

Catalog No. 22-046-792

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

  • Measurement range of 0–4000 mOsm/kg H2O
  • Features 0.2 to 0.25mL sample volume
  • Frost-free cooling chamber eliminates most routine maintenance
  • Freezing-point method to accurately determine total concentration
  • On-board sample identification and broad measurement range
  • Results in two minutes for a wide variety of laboratory applications
  • Intel™ microprocessor
  • On-board printer or connect to external printer, computer and bar code scanner
  • Stores up to 30 test results
  • Other features: push-button design, multi-language capability, statistical analysis and automated calibration

Specifications

Specifications

1 Year
2A
250V
18 in.
Each
3250
Ideal for serum, urine or other bodily fluids in clinical applications, media or formulations in pharmaceutical, biotech or industrial applications
12 in.
30.5cm
≈2 min. per sample in low range, ≈3 min. in high range
0 to 2000mOsm/kg H2O
1mOsm/kg H2O
-40° to 45°C
18° to 35°C
23 lb.
33cm
250V 50/60Hz
Healthcare
50/60Hz
CE, ETD, IVD
45.7cm
Single Sample Osmometer
Vacuum Fluorescent Display (VFD)
<1mOsm/kg H2O
<0.5%
5 to 80%
100 to 250VAC
0 to 400mOsm: ±2mOsm/kg (1 S.D.), 400 to 4000mOsm: ±0.5% (1 S.D.)
200 to 250μL
64° to 95°F
95w
10.4kg
13 in.
Documents