Please login to your online account to display your discounted pricing

Advanced Instruments Renol™ Urine Control Solution

$270.74 - $701.06

Specifications

Certifications/Compliance CE, CLIA, CAP
For Use With (Application) Evaluating the performance the osmometer in clinical laboratories testing Human Urine
Storage Requirements 2° to 8°C
View More Specs
Products
Catalog Number Mfr. No. Concentration Shelf Life Price Quantity    

22-046-221

 
advanced instruments inc
3LA085
300 and 800mOsm/kg H2O Unopened: 2 Years and Opened: 10 days when refrigerated Pack of 8 Each for $701.06

22-046-607

 
advanced instruments inc
200219*
800mOsm/kg H2O Unopened: 2 Years; Opened: 10 days when refrigerated Pack of 8 Each for $270.74

22-046-606

 
advanced instruments inc
200218
300mOsm/kg H2O Unopened: 2 Years; Opened: 10 days when refrigerated Pack of 8 Each for $270.74

22-046-605

 
advanced instruments inc
200217
300 and 800mOsm/kg H2O Unopened: 2 Years; Opened: 10 days when refrigerated Pack of 8 Each for $270.74
Description & Specifications

Specifications

Certifications/Compliance CE, CLIA, CAP
For Use With (Application) Evaluating the performance the osmometer in clinical laboratories testing Human Urine
Storage Requirements 2° to 8°C

  • For use with Advanced Osmometers: 3320, 2020, 3250, 3900, and 210

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.

Safety and Handling

Unopened: 2 Years and Opened: 10 days when refrigerated