Please login to your online account to display your discounted pricing

Advanced Instruments Protinol™ Serum Osmometer Controls

Quality control check for the clinical laboratory for osmometers from any manufacturer

$285.66 - $692.89

Specifications

Certifications/Compliance NIST, ISO 9000, CLIA, CAP
For Use With (Application) Use in evaluating performance of osmometer in clinical laboratories testing of Human Serum samples
Shelf Life Unopened: 18 months; Opened: 7 days when refrigerated
Storage Requirements 2° to 8°C
View More Specs
Products
Catalog Number Mfr. No. Concentration Quantity Price Quantity    

22-046781

 
advanced instruments inc
3MA028
240, 280, 320mOsm/kg H2O 9 Pack Each for $692.89

22-046-601

 
advanced instruments inc
200213
240, 280, 320mOsm/kg H2O 12 Pack Pack of 12 Each for $428.49

22-046-602

 
advanced instruments inc
200214
240mOsm/kg H2O 8 Pack Pack of 8 Each for $285.66

22-046-603

 
advanced instruments inc
200215
280mOsm/kg H2O 8 Pack Pack of 8 Each for $285.66

22-046-604

 
advanced instruments inc
200216
320mOsm/kg H2O 8 Pack Pack of 8 Each for $285.66
Description & Specifications

Specifications

Certifications/Compliance NIST, ISO 9000, CLIA, CAP
For Use With (Application) Use in evaluating performance of osmometer in clinical laboratories testing of Human Serum samples
Shelf Life Unopened: 18 months; Opened: 7 days when refrigerated
Storage Requirements 2° to 8°C

  • Developed from bovine serum albumin — adjusted with nonprotein material
  • Produce consistent, reliable results in the human serum range
  • Run controls daily to check and document the osmometer's performance
  • Set of three values: 240 ±5mOsm/kg H2O (abnormal low), 280 ±5mOsm/kg H2O (normal), and 320 ±5mOsm/kg H2O (abnormal high)

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.

Safety and Handling

Unopened: 18 months; Opened: 7 days when refrigerated