Please login to your online account to display your discounted pricing

Alfa Aesar™ Adrenorphin, Free acid

For cellular and molecular biology applications

Specifications

Product Type Adrenorphin, free acid
Molecular Weight 985.16
Storage Requirements -30° to -10°C
Sequence Tyr-Gly-Gly-Phe-Met-Arg-Arg-Val
Format Solid
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66389LB0

 
alfa aesar™
J66389LB0
5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66389MCR

 
alfa aesar™
J66389MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Adrenorphin, free acid
Molecular Weight 985.16
Storage Requirements -30° to -10°C
Sequence Tyr-Gly-Gly-Phe-Met-Arg-Arg-Val
Format Solid
Regulatory Status RUO
Purification Method HPLC