Please login to your online account to display your discounted pricing

Alfa Aesar™ Adrenomedullin (16-31), Human, Pig

For cellular and molecular biology applications

Specifications

Product Type Adrenomedullin (16-31), human, pig
Cross Reactivity Human, Pig
Molecular Weight 1865.18
Storage Requirements -30° to -10°C
Format Solid
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66935EXD

 
alfa aesar™
J66935EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66935MCR

 
alfa aesar™
J66935MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Adrenomedullin (16-31), human, pig
Cross Reactivity Human, Pig
Molecular Weight 1865.18
Storage Requirements -30° to -10°C
Format Solid
Sequence Cys-Arg-Phe-Gly-Thr-Cys-Thr-Val-Gln-Lys-Leu-Ala-His-Gln-Ile-Tyr-NH2 (Disulfide bridge Cys16-Cys21)
Regulatory Status RUO
Purification Method HPLC