Please login to your online account to display your discounted pricing

Alfa Aesar™ Adrenomedullin (1-52), Porcine

For cellular and molecular biology applications

Specifications

Product Type Adrenomedullin (1-52), porcine
Cross Reactivity Porcine
Molecular Weight 5971.67
Storage Requirements -30° to -10°C
Format Solid
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66356EXD

 
alfa aesar™
J66356EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66356MCR

 
alfa aesar™
J66356MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type Adrenomedullin (1-52), porcine
Cross Reactivity Porcine
Molecular Weight 5971.67
Storage Requirements -30° to -10°C
Format Solid
Regulatory Status RUO
Purification Method HPLC