missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Adrenocorticotropic Hormone (4-11), human, Alfa Aesar™

$70.24 - $173.58

Specifications

Format Solid
CAS 67224-41-3
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAJ66357MCR
View Documents
Thermo Fisher Scientific
J66357MCR
1mg Each for $70.24
 
AAJ66357LB0
View Documents
Thermo Fisher Scientific
J66357LB0
5mg Each for $173.58
 
Specifications

Specifications

Solid
67224-41-3