Please login to your online account to display your discounted pricing

Thermo Scientific™ Lab Vision™ Adrenocorticotrophic Hormone (ACTH), Rabbit Polyclonal Antibody

Obtain accurate, reproducible results in western blotting experiments with Thermo Scientific™ Adrenocorticotrophic Hormone (ACTH), Rabbit Polyclonal Antibody.

$212.49 - $1,042.86

Specifications

Antigen Adrenocorticotrophic Hormone (ACTH)
Concentration 200µg/mL
Applications Western Blotting
Classification Polyclonal
Conjugate Unlabeled
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

RB9217P

 
thermo scientific
RB9217P
1mL Each for $1,042.86

RB9217P0

 
thermo scientific™ lab vision™
RB9217P0
100µL Each for $212.49

RB9217P1

 
thermo scientific™ lab vision™
RB9217P1
500µL Each for $632.02
Description & Specifications

Specifications

Antigen Adrenocorticotrophic Hormone (ACTH)
Concentration 200µg/mL
Applications Western Blotting
Classification Polyclonal
Conjugate Unlabeled
Cross Reactivity Human
Format Concentrated
Host Species Rabbit
Regulatory Status RUO
Research Discipline Steroids Hormones and Growth Factors
Type Primary
Immunogen Synthetic peptide derived from N-terminus of human ACTH
Isotype IgG

Obtain accurate, reproducible results in western blotting experiments with Thermo Scientific™ Adrenocorticotrophic Hormone (ACTH), Rabbit Polyclonal Antibody.

ACTH labels corticotrophs in the adenohypophysis and is useful in the classification of pituitary adenomas.

Host Species: Rabbit

Isotype: IgG

Species Reactivity: Human. It shows broad species reactivity.

Epitope: aa 1-24

Immunogen: Synthetic peptide derived from N-terminus of human ACTH

Positive Control: ACTH recombinant protein

Cellular Localization: Cytoplasmic

Recommended for:

  • Western Blotting

Note:

USA: RUO; Int'l: RUO